siRNA Details
| Gene Symbol: TTC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031421.3 | NP_113609.1 | 10 | 1465 | ||
| NR_110662.1 | 8 | 1032 | ||||
| XM_011525323.2 | XP_011523625.1 | 10 | 1405 | |||
| GenBank mRNA | AK055498.1 | - | 10 | 1410 | ||
| AL136760.1 | - | 10 | 1432 | |||
| BC025390.1 | - | 8 | 1040 | |||
| BC067297.1 | - | 10 | 1424 | 30383608 | ||

TTC25
Genomic Map