siRNA Details
| Gene Symbol: TTC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031421.3 | NP_113609.1 | 6 | 861 | ||
| XM_011525323.2 | XP_011523625.1 | 6 | 801 | |||
| XM_011525324.2 | XP_011523626.1 | 6 | 828 | |||
| GenBank mRNA | AK055498.1 | - | 6 | 806 | ||
| AL136760.1 | - | 6 | 828 | |||
| BC067297.1 | - | 6 | 820 | 30383608 | ||

TTC25
Genomic Map