siRNA Details
| Gene Symbol: RILP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031430.2 | NP_113618.2 | 5 | 1057 | ||
| XM_005256811.1 | XP_005256868.1 | 5 | 1036 | |||
| GenBank mRNA | AF370391.1 | - | 5 | 994 | ||
| AJ278711.1 | - | 5 | 1057 | |||
| AJ404317.1 | - | 5 | 1027 | |||
| AK055755.1 | - | 5 | 1036 | |||
| AK314767.1 | - | 5 | 1036 | |||
| BC004961.1 | - | 5 | 606 | |||
| BC031621.1 | - | 5 | 851 | 5174974 | ||

RILP
Genomic Map