siRNA Details
| Gene Symbol: RILP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031430.2 | NP_113618.2 | 7 | 1266 | ||
| GenBank mRNA | AF370391.1 | - | 7 | 1203 | ||
| AJ278711.1 | - | 7 | 1266 | |||
| AJ404317.1 | - | 7 | 1236 | |||
| AK055755.1 | - | 7 | 1245 | |||
| AK314767.1 | - | 7 | 1245 | |||
| BC004961.1 | - | 7 | 815 | |||
| BC031621.1 | - | 7 | 1060 | 5174974 | ||

RILP
Genomic Map