siRNA Details
| Gene Symbol: RILP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031430.2 | NP_113618.2 | 8 | 1351 | ||
| XM_005256811.1 | XP_005256868.1 | 7 | 1246 | |||
| XM_017025195.1 | XP_016880684.1 | 6 | 1100 | |||
| GenBank mRNA | AF370391.1 | - | 8 | 1288 | ||
| AJ278711.1 | - | 8 | 1351 | |||
| AJ404317.1 | - | 8 | 1321 | |||
| AK055755.1 | - | 8 | 1330 | |||
| AK314767.1 | - | 8 | 1330 | |||
| BC004961.1 | - | 8 | 900 | |||
| BC031621.1 | - | 8 | 1145 | 5174974 | ||

RILP
Genomic Map