siRNA Details
| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031438.3 | NP_113626.1 | 2 | 285 | ||
| XM_005272095.1 | XP_005272152.1 | 2 | 270 | |||
| GenBank mRNA | AK098066.1 | - | 2 | 254 | ||
| AK309942.1 | - | 2 | 270 | |||
| AL136592.1 | - | 2 | 240 | |||
| BC041099.1 | - | 2 | 269 | 5495144 | ||
| CB131118.1 | - | 2 | 273 | |||
| DB029218.1 | - | 2 | 287 | |||

NUDT12
Genomic Map