siRNA Details
| Gene Symbol: CCDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282658.1 | NP_001269587.1 | 6 | 1216 | ||
| NM_031455.3 | NP_113643.1 | 2 | 651 | |||
| GenBank mRNA | AK095792.1 | - | 6 | 1095 | ||
| AL136562.1 | - | 2 | 567 | |||
| BC051334.1 | - | 2 | 651 | 6302570 | ||

CCDC3
Genomic Map