siRNA Details
| Gene Symbol: CRISPLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286777.1 | NP_001273706.1 | 4 | 560 | ||
| NM_001286778.1 | NP_001273707.1 | 5 | 676 | |||
| NM_031461.5 | NP_113649.1 | 6 | 1188 | |||
| GenBank mRNA | AF142573.1 | - | 6 | 784 | ||
| AF329197.1 | - | 5 | 763 | |||
| AK304034.1 | - | 5 | 664 | |||
| AK304350.1 | - | 4 | 541 | |||
| AK314048.1 | - | 6 | 944 | |||
| AL834301.1 | - | 6 | 920 | |||
| AY358564.1 | - | 6 | 1146 | |||
| BC020514.1 | - | 6 | 1053 | 3856615 | ||

CRISPLD1
Genomic Map