siRNA Details
| Gene Symbol: JAM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205329.1 | NP_001192258.1 | 7 | 873 | ||
| NM_032801.4 | NP_116190.3 | 8 | 1026 | |||
| GenBank mRNA | AF356518.1 | - | 8 | 867 | ||
| AF448478.1 | - | 8 | 901 | |||
| AJ344431.1 | - | 8 | 882 | |||
| AJ416101.1 | - | 8 | 1026 | |||
| AK074769.1 | - | 8 | 909 | |||
| AK075309.1 | - | 8 | 880 | |||
| AK125071.1 | - | 7 | 727 | |||
| AY358335.1 | - | 8 | 917 | |||
| BC010690.1 | - | 8 | 865 | |||
| BC012147.1 | - | 8 | 867 | 4552285 | ||
| BC057284.1 | - | 8 | 880 | |||

JAM3
Genomic Map