siRNA Details
| Gene Symbol: YPEL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145524.1 | NP_001138996.1 | 2 | 202 | ||
| NM_031477.4 | NP_113665.3 | 1 | 848 | |||
| GenBank mRNA | AB098737.1 | - | 2 | 202 | ||
| AF305622.1 | - | 2 | 205 | |||
| AK022409.1 | - | 1 | 1071 | |||
| AK026047.1 | - | 1 | 236 | |||
| BC005009.1 | - | 2 | 158 | 3639517 | ||
| BC050664.1 | - | 1 | 847 | 5748830 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026875, NM_025347, AK002925, AF523353, AK004431, and others.

YPEL3
Genomic Map