siRNA Details
Gene Symbol: SYT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031914.2 | NP_114120.2 | 4 | 1611 | ||
XM_006720272.3 | XP_006720335.1 | 6 | 2071 | |||
XM_011537219.1 | XP_011535521.1 | 6 | 2148 | |||
XM_011537220.1 | XP_011535522.1 | 6 | 2148 | |||
XM_011537221.2 | XP_011535523.1 | 6 | 2148 | |||
XM_011537222.2 | XP_011535524.1 | 6 | 2148 | |||
XM_011537223.2 | XP_011535525.1 | 5 | 1185 | |||
XM_011537224.2 | XP_011535526.1 | 6 | 1331 | |||
XM_011537225.2 | XP_011535527.1 | 6 | 1630 | |||
XM_011537227.2 | XP_011535529.2 | 4 | 1008 | |||
XM_011537228.2 | XP_011535530.1 | 4 | 951 | |||
XM_011537229.2 | XP_011535531.1 | 3 | 795 | |||
XM_017021693.1 | XP_016877182.1 | 6 | 2148 | |||
XM_017021694.1 | XP_016877183.1 | 6 | 1837 | |||
XM_017021695.1 | XP_016877184.1 | 7 | 1982 | |||
XM_017021696.1 | XP_016877185.1 | 5 | 1130 | |||
XM_017021697.1 | XP_016877186.1 | 5 | 1138 | |||
XM_017021698.1 | XP_016877187.1 | 5 | 1254 | |||
GenBank mRNA | AB102949.1 | - | 4 | 1354 | ||
AJ617628.1 | - | 4 | 1612 | |||
BC040924.1 | - | 2 | 503 | |||
BC117363.1 | - | 4 | 1470 | 40125914 | ||
BC143598.1 | - | 4 | 1470 | 9052110 |
