siRNA Details
| Gene Symbol: TMEM120A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031925.2 | NP_114131.1 | 12 | 1089 | ||
| GenBank mRNA | AF327923.1 | - | 12 | 1050 | ||
| BC029487.1 | - | 10 | 1208 | 5277668 | ||
| BC051850.1 | - | 12 | 1074 | 6201334 | ||
| CX869634.1 | - | 1 | 405 | |||

TMEM120A
Genomic Map