siRNA Details
| Gene Symbol: HMCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031935.2 | NP_114141.2 | 85, 86 | 13447 | ||
| XM_011510038.2 | XP_011508340.1 | 85, 86 | 13464 | |||
| XM_017002437.1 | XP_016857926.1 | 74, 75 | 11397 | |||
| GenBank mRNA | AF156100.1 | - | 85, 86 | 13450 | ||
| AJ306906.1 | - | 28, 29 | 4332 | |||
| AL833232.1 | - | 3, 4 | 396 | |||

HMCN1
Genomic Map