siRNA Details
| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031936.4 | NP_114142.3 | 2 | 1096 | ||
| XM_017002438.1 | XP_016857927.1 | 2 | 3429 | |||
| GenBank mRNA | AF258342.1 | - | 3 | 732 | ||
| AF317652.1 | - | 1 | 413 | |||
| AK290027.1 | - | 2 | 1096 | |||
| BC067464.1 | - | 1 | 481 | 6971762 | ||
| JF810889.1 | - | 1 | 413 | |||
More Information
Set Membership: |
|

GPR61
Genomic Map