siRNA Details
| Gene Symbol: TSPAN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290212.1 | NP_001277141.1 | 3 | 570 | ||
| NM_031945.4 | NP_114151.3 | 2 | 336 | |||
| NR_110866.1 | 2 | 336 | ||||
| GenBank mRNA | AB593081.1 | - | 3 | 543 | ||
| AB593082.1 | - | 2 | 331 | |||
| AF325213.1 | - | 2 | 292 | |||
| AK096926.1 | - | 2 | 318 | |||
| BC021923.1 | - | 2 | 305 | 4876887 | ||
| BC032802.1 | - | 2 | 307 | 5191718 | ||
| BG760374.1 | - | 2 | 305 | |||
| BM710905.1 | - | 2 | 338 | |||
| DA912496.1 | - | 2 | 318 | |||

TSPAN10
Genomic Map