siRNA Details
| Gene Symbol: FGFBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031950.3 | NP_114156.1 | 1 | 709 | ||
| GenBank mRNA | AB021123.1 | - | 1 | 680 | ||
| AF361936.1 | - | 1 | 602 | |||
| AY358978.1 | - | 1 | 709 | |||
| BC025720.1 | - | 1 | 678 | 5228917 | ||
| CN480296.1 | - | 2 | 476 | |||

FGFBP2
Genomic Map