siRNA Details
| Gene Symbol: GSG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031965.2 | NP_114171.2 | 1 | 815 | ||
| GenBank mRNA | AB039834.3 | - | 1 | 812 | ||
| AF289865.1 | - | 1 | 811 | |||
| AK056691.1 | - | 1 | 816 | |||
| AK292715.1 | - | 1 | 801 | |||
| AK310546.1 | - | 1 | 815 | |||
| AK315247.1 | - | 1 | 815 | |||
| BC016626.1 | - | 1 | 801 | |||
| BC047457.1 | - | 1 | 796 | 5298590 | ||

GSG2
Genomic Map