siRNA Details
| Gene Symbol: SPRTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010984.3 | NP_001010984.1 | 1 | 584 | ||
| NM_001261462.2 | NP_001248391.1 | 1 | 584 | |||
| NM_032018.6 | NP_114407.3 | 1 | 584 | |||
| XM_006711818.2 | XP_006711881.1 | 1 | 556 | |||
| GenBank mRNA | AK027317.1 | - | 1 | 255 | ||
| AK027613.1 | - | 1 | 264 | |||
| AL512744.1 | - | 1 | 198 | |||
| AY358611.1 | - | 1 | 1417 | |||
| BC015740.1 | - | 1 | 472 | |||
| BC068478.1 | - | 1 | 535 | 5298048 | ||

SPRTN
Genomic Map