siRNA Details
| Gene Symbol: SPRTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010984.3 | NP_001010984.1 | 4 | 1016 | ||
| NM_001261462.2 | NP_001248391.1 | 3 | 887 | |||
| NM_032018.6 | NP_114407.3 | 4 | 1016 | |||
| XM_006711818.2 | XP_006711881.1 | 3 | 859 | |||
| XM_011544289.2 | XP_011542591.1 | 2 | 342 | |||
| GenBank mRNA | AK027317.1 | - | 3 | 558 | ||
| AK027613.1 | - | 4 | 696 | |||
| AL512744.1 | - | 4 | 630 | |||
| AY358611.1 | - | 4 | 1849 | |||
| BC015740.1 | - | 4 | 904 | |||
| BC068478.1 | - | 4 | 967 | 5298048 | ||

SPRTN
Genomic Map