siRNA Details
Gene Symbol: CTTNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033427.2 | NP_219499.1 | 13 | 3482 | ||
XM_005250634.4 | XP_005250691.1 | 13 | 3617 | |||
XM_005250635.3 | XP_005250692.1 | 13 | 3251 | |||
XM_011516615.2 | XP_011514917.1 | 13 | 5785 | |||
XM_011516616.2 | XP_011514918.1 | 13 | 3414 | |||
XM_017012706.1 | XP_016868195.1 | 13 | 3414 | |||
XM_017012708.1 | XP_016868197.1 | 10 | 2996 | |||
GenBank mRNA | AB051545.1 | - | 13 | 3388 | ||
AF377960.1 | - | 13 | 3482 | |||
AK096565.1 | - | 1 | 19 | |||
BC106000.1 | - | 13 | 3449 | 30528261 |
