siRNA Details
| Gene Symbol: MND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253861.1 | NP_001240790.1 | 4, 5 | 370 | ||
| NM_032117.3 | NP_115493.1 | 4, 5 | 370 | |||
| XM_005263275.2 | XP_005263332.1 | 4, 5 | 329 | |||
| XM_017008688.1 | XP_016864177.1 | 4, 5 | 727 | |||
| GenBank mRNA | AK312172.1 | - | 4, 5 | 329 | ||
| AY028916.1 | - | 4, 5 | 309 | |||
| BC032142.2 | - | 4, 5 | 324 | 5095005 | ||
| BG613292.1 | - | 4, 5 | 332 | |||
| BU533354.1 | - | 1, 2 | 26 | |||

MND1
Genomic Map