siRNA Details
| Gene Symbol: PLEKHN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160184.1 | NP_001153656.1 | 12 | 1269 | ||
| NM_032129.2 | NP_115505.2 | 13 | 1374 | |||
| XM_006710944.3 | XP_006711007.2 | 13 | 2087 | |||
| XM_011542248.2 | XP_011540550.2 | 12 | 2207 | |||
| XM_017002474.1 | XP_016857963.1 | 11 | 2066 | |||
| XM_017002478.1 | XP_016857967.1 | 11 | 2032 | |||
| GenBank mRNA | AL136730.1 | - | 13 | 1374 | ||
| BC101386.2 | - | 12 | 1264 | 40026400 | ||
| BC101387.2 | - | 12 | 1525 | 40026404 | ||

PLEKHN1
Genomic Map