siRNA Details
| Gene Symbol: ZRANB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286568.1 | NP_001273497.1 | 15 | 2395 | ||
| NM_001286569.1 | NP_001273498.1 | 16 | 2524 | |||
| NM_032143.3 | NP_115519.2 | 15 | 2306 | |||
| XM_005263809.1 | XP_005263866.1 | 15 | 2260 | |||
| XM_006712788.1 | XP_006712851.1 | 15 | 2234 | |||
| XM_011511958.2 | XP_011510260.1 | 15 | 2749 | |||
| XM_011511959.2 | XP_011510261.2 | 15 | 2314 | |||
| XM_011511960.1 | XP_011510262.1 | 15 | 2282 | |||
| XM_011511961.2 | XP_011510263.2 | 15 | 2248 | |||
| XM_011511963.1 | XP_011510265.1 | 15 | 2247 | |||
| XM_011511966.2 | XP_011510268.2 | 13 | 2132 | |||
| XM_017005065.1 | XP_016860554.1 | 15 | 2253 | |||
| XM_017005066.1 | XP_016860555.1 | 15 | 2312 | |||
| XM_017005067.1 | XP_016860556.1 | 15 | 2260 | |||
| XM_017005068.1 | XP_016860557.1 | 15 | 2234 | |||
| XM_017005069.1 | XP_016860558.1 | 15 | 2282 | |||
| XM_017005070.1 | XP_016860559.1 | 15 | 2276 | |||
| XM_017005071.1 | XP_016860560.1 | 13 | 2053 | |||
| GenBank mRNA | AK095362.1 | - | 14 | 1964 | ||
| AK131303.1 | - | 9 | 1450 | |||
| AK304601.1 | - | 3 | 641 | |||
| AL136824.1 | - | 4 | 742 | |||
| AL833322.1 | - | 16 | 2405 | |||
| BC089429.1 | - | 15 | 2306 | 3077975 | ||
| BX647838.1 | - | 15 | 2423 | |||

ZRANB3
Genomic Map