siRNA Details
| Gene Symbol: SNX29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032167.4 | NP_115543.3 | 1, 2 | 86 | ||
| XM_017023872.1 | XP_016879361.1 | 1, 2 | 75 | |||
| XM_017023874.1 | XP_016879363.1 | 1, 2 | 75 | |||
| XM_017023877.1 | XP_016879366.1 | 1, 2 | 75 | |||
| GenBank mRNA | AK022425.1 | - | 1, 2 | 75 | ||
| AL135642.1 | - | 1, 2 | 75 | |||
| HY078299.1 | - | 1, 2 | 87 | |||

SNX29
Genomic Map