siRNA Details
| Gene Symbol: EVA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135032.1 | NP_001128504.1 | 3, 4 | 552 | ||
| NM_032181.2 | NP_115557.1 | 3, 4 | 512 | |||
| GenBank mRNA | AF177337.1 | - | 586 | |||
| AK023453.1 | - | 3, 4 | 512 | |||
| BC016157.1 | - | 3, 4 | 488 | 2989786 | ||
| BC063016.1 | - | 5, 6 | 618 | 5226973 | ||
| BE892427.1 | - | 3, 4 | 503 | |||
| BP276065.1 | - | 552 | ||||
| CR457314.1 | - | 1, 2 | 74 | |||

EVA1A
Genomic Map