siRNA Details
| Gene Symbol: MFSD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294341.1 | NP_001281270.1 | 4, 5 | 901 | ||
| NM_032219.3 | NP_115595.2 | 4, 5 | 901 | |||
| XM_005272299.4 | XP_005272356.1 | 4, 5 | 928 | |||
| XM_006713918.1 | XP_006713981.1 | 4, 5 | 555 | |||
| XM_011513568.1 | XP_011511870.1 | 4, 5 | 1705 | |||
| XM_011513571.1 | XP_011511873.1 | 5, 6 | 725 | |||
| XM_011513573.2 | XP_011511875.1 | 4, 5 | 928 | |||
| XM_017008700.1 | XP_016864189.1 | 4, 5 | 928 | |||
| GenBank mRNA | AK292010.1 | - | 4, 5 | 799 | ||
| AY358401.1 | - | 4, 5 | 603 | |||
| BC030246.1 | - | 4, 5 | 644 | 5216645 | ||

MFSD7
Genomic Map