siRNA Details
| Gene Symbol: FAM96A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014812.2 | NP_001014812.1 | 2 | 419 | ||
| NM_001289108.1 | NP_001276037.1 | 2 | 419 | |||
| NM_032231.5 | NP_115607.1 | 2 | 419 | |||
| NR_110310.1 | 3 | 537 | ||||
| GenBank mRNA | AK304202.1 | - | 2 | 206 | ||
| AK312171.1 | - | 2 | 361 | |||
| BC008865.2 | - | 2 | 321 | 4128736 | ||
| BM706342.1 | - | 2 | 419 | |||
| BQ269528.1 | - | 2 | 407 | |||
| CN421575.1 | - | 2 | 347 | |||
| DC415981.1 | - | 3 | 461 | |||

FAM96A
Genomic Map