siRNA Details
| Gene Symbol: TMEM117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286211.1 | NP_001273140.1 | 2, 3 | 646 | ||
| NM_001286212.1 | NP_001273141.1 | 2, 3 | 646 | |||
| NM_032256.2 | NP_115632.1 | 2, 3 | 646 | |||
| XM_011538831.2 | XP_011537133.1 | 2, 3 | 655 | |||
| XM_011538832.2 | XP_011537134.1 | 2, 3 | 330 | |||
| XM_017020023.1 | XP_016875512.1 | 2, 3 | 372 | |||
| GenBank mRNA | AK057287.1 | - | 2, 3 | 365 | ||
| AL136855.1 | - | 2, 3 | 348 | |||
| BC030284.1 | - | 2, 3 | 354 | 5240522 | ||
| BC060798.1 | - | 2, 3 | 409 | 30348706 | ||

TMEM117
Genomic Map