siRNA Details
| Gene Symbol: DRC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289162.1 | NP_001276091.1 | 7 | 953 | ||
| NM_001289163.1 | NP_001276092.1 | 5 | 627 | |||
| NM_032269.5 | NP_115645.4 | 6 | 822 | |||
| XM_011523377.1 | XP_011521679.1 | 6 | 828 | |||
| XM_011523378.1 | XP_011521680.1 | 5 | 633 | |||
| GenBank mRNA | AK093035.1 | - | 5 | 592 | ||
| AK292558.1 | - | 6 | 817 | |||
| BC030661.1 | - | 7 | 947 | |||
| BC036667.1 | - | 6 | 812 | 4828649 | ||

DRC7
Genomic Map