siRNA Details
| Gene Symbol: TMEM126A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244735.1 | NP_001231664.1 | 3, 4 | 474 | ||
| NM_032273.3 | NP_115649.1 | 4, 5 | 567 | |||
| GenBank mRNA | AK312081.1 | - | 4, 5 | 499 | ||
| AL136941.1 | - | 4, 5 | 513 | |||
| BC007875.2 | - | 4, 5 | 485 | 3948518 | ||
| BQ943238.1 | - | 4, 5 | 511 | |||
| CD366181.1 | - | 1, 2 | 248 | |||

TMEM126A
Genomic Map