siRNA Details
| Gene Symbol: TMEM126A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244735.1 | NP_001231664.1 | 4 | 519 | ||
| NM_032273.3 | NP_115649.1 | 5 | 612 | |||
| GenBank mRNA | AK312081.1 | - | 5 | 544 | ||
| AL136941.1 | - | 5 | 558 | |||
| BC007875.2 | - | 5 | 530 | 3948518 | ||
| BQ943238.1 | - | 5 | 556 | |||
| CD366181.1 | - | 1 | 203 | |||

TMEM126A
Genomic Map