siRNA Details
Gene Symbol: PSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032289.2 | NP_115665.1 | 5 | 1258 | ||
XM_011537696.2 | XP_011535998.1 | 6 | 1314 | |||
XM_017009975.1 | XP_016865464.1 | 6 | 2621 | |||
XM_017009976.1 | XP_016865465.1 | 6 | 3429 | |||
XM_017009977.1 | XP_016865466.1 | 6 | 3309 | |||
GenBank mRNA | AL136559.1 | - | 5 | 1258 | ||
AL834268.1 | - | 1 | 40 | |||
BC038233.1 | - | 5 | 1235 | 5752229 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001289602, NM_028707, BC065055, AK053835, AK034575, and others.
Panther Classification:
Gene Ontology Categories:
