siRNA Details
| Gene Symbol: NICN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032316.3 | NP_115692.1 | 5, 6 | 686 | ||
| GenBank mRNA | AF538150.1 | - | 4, 5 | 950 | ||
| AJ299740.1 | - | 5, 6 | 658 | |||
| AK094248.1 | - | 5, 6 | 626 | |||
| AK301846.1 | - | 4, 5 | 575 | |||
| AK313073.1 | - | 5, 6 | 683 | |||
| BC005050.1 | - | 5, 6 | 603 | 2820022 | ||
| BC050653.1 | - | 5, 6 | 650 | 6015080 | ||

NICN1
Genomic Map