siRNA Details
| Gene Symbol: TMEM246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303107.1 | NP_001290036.1 | 3 | 1502 | ||
| NM_001303108.1 | NP_001290037.1 | 2 | 1377 | |||
| NM_032342.2 | NP_115718.1 | 2 | 1447 | |||
| XM_017015206.1 | XP_016870695.1 | 2 | 1637 | |||
| GenBank mRNA | AK313981.1 | - | 2 | 1358 | ||
| BC006115.1 | - | 2 | 1419 | 3503592 | ||
| BC033550.1 | - | 4 | 2245 | 4822472 | ||
| BC113967.1 | - | 1 | 624 | |||

TMEM246
Genomic Map