siRNA Details
| Gene Symbol: TMEM246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303107.1 | NP_001290036.1 | 3 | 482 | ||
| NM_001303108.1 | NP_001290037.1 | 2 | 357 | |||
| NM_032342.2 | NP_115718.1 | 2 | 427 | |||
| XM_017015206.1 | XP_016870695.1 | 2 | 617 | |||
| GenBank mRNA | AL521069.3 | - | 3 | 386 | ||
| BC006115.1 | - | 2 | 399 | 3503592 | ||
| BC033550.1 | - | 4 | 1225 | 4822472 | ||
| HY033877.1 | - | 2 | 358 | |||
| HY174800.1 | - | 2 | 428 | |||

TMEM246
Genomic Map