siRNA Details
| Gene Symbol: PYM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143853.1 | NP_001137325.1 | 3 | 355 | ||
| NM_032345.2 | NP_115721.1 | 3 | 494 | |||
| GenBank mRNA | AJ459406.1 | - | 3 | 342 | ||
| AK096922.1 | - | 3 | 342 | |||
| BC006135.1 | - | 3 | 494 | 3640407 | ||
| BC014976.1 | - | 3 | 463 | 4854916 | ||
| DB466388.1 | - | 3 | 356 | |||

PYM1
Genomic Map