siRNA Details
| Gene Symbol: SARNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033082.3 | NP_149073.1 | 7, 8 | 471 | ||
| NR_026722.1 | 6, 7 | 424 | ||||
| NR_026723.1 | 7, 8 | 471 | ||||
| GenBank mRNA | AF486281.1 | - | 7, 8 | 416 | ||
| AK290508.1 | - | 7, 8 | 439 | |||
| BC007099.1 | - | 7, 8 | 437 | 4273903 | ||
| BC093051.1 | - | 6, 7 | 390 | 30382893 | ||
| BI520421.1 | - | 7, 8 | 471 | |||

SARNP
Genomic Map