siRNA Details
| Gene Symbol: ZNF414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146175.1 | NP_001139647.1 | 5 | 890 | ||
| NM_032370.2 | NP_115746.2 | 5 | 890 | |||
| GenBank mRNA | AI937020.1 | - | 2 | 311 | ||
| AK074191.1 | - | 5 | 886 | |||
| AK225034.1 | - | 5 | 886 | |||
| BC007310.1 | - | 5 | 874 | 3353224 | ||
| BC071937.1 | - | 5 | 869 | 5089259 | ||
| BQ773835.1 | - | 4 | 527 | |||
| BU621653.1 | - | 4 | 528 | |||

ZNF414
Genomic Map