siRNA Details
| Gene Symbol: PCGF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256549.1 | NP_001243478.1 | 8 | 975 | ||
| NM_001257101.1 | NP_001244030.1 | 8 | 868 | |||
| NM_032373.4 | NP_115749.2 | 8 | 1033 | |||
| XM_006718030.1 | XP_006718093.1 | 7 | 964 | |||
| XM_011540271.2 | XP_011538573.1 | 8 | 1034 | |||
| XM_011540272.2 | XP_011538574.1 | 8 | 1065 | |||
| XM_017016776.1 | XP_016872265.1 | 8 | 980 | |||
| XM_017016777.1 | XP_016872266.1 | 8 | 979 | |||
| XM_017016778.1 | XP_016872267.1 | 7 | 964 | |||
| GenBank mRNA | AK303014.1 | - | 8 | 976 | ||
| AL832003.2 | - | 6 | 472 | |||
| AL832496.1 | - | 8 | 849 | |||
| BC051845.1 | - | 8 | 843 | 5199133 | ||

PCGF5
Genomic Map