siRNA Details
| Gene Symbol: C2orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032411.2 | NP_115787.1 | 1, 2 | 193 | ||
| GenBank mRNA | AF325503.1 | - | 1, 2 | 193 | ||
| AY358601.1 | - | 1, 2 | 154 | |||
| BC021742.1 | - | 1, 2 | 197 | 5260075 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf40| Gene Symbol: C2orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032411.2 | NP_115787.1 | 1, 2 | 193 | ||
| GenBank mRNA | AF325503.1 | - | 1, 2 | 193 | ||
| AY358601.1 | - | 1, 2 | 154 | |||
| BC021742.1 | - | 1, 2 | 197 | 5260075 | ||