siRNA Details
| Gene Symbol: CARD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324281.1 | NP_001311210.1 | 18 | 2650 | ||
| NM_032415.5 | NP_115791.3 | 17 | 2589 | |||
| XM_011515586.2 | XP_011513888.1 | 18 | 2618 | |||
| XM_011515587.2 | XP_011513889.1 | 17 | 2581 | |||
| GenBank mRNA | AF322641.1 | - | 17 | 2491 | ||
| AF352576.1 | - | 15 | 2164 | |||
| AK074049.1 | - | 17 | 2509 | |||
| BC111719.1 | - | 16 | 2275 | 40009168 | ||

CARD11
Genomic Map