siRNA Details
| Gene Symbol: BRSK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032430.1 | NP_115806.1 | 4 | 633 | ||
| XM_005259327.3 | XP_005259384.1 | 4 | 702 | |||
| XM_011527395.2 | XP_011525697.2 | 2 | 67 | |||
| GenBank mRNA | AB058714.1 | - | 3 | 168 | ||
| AF479826.1 | - | 6 | 531 | |||
| AF479827.1 | - | 4 | 633 | |||
| AL834275.1 | - | 4 | 256 | |||
| AY458602.1 | - | 4 | 405 | |||
| AY505124.1 | - | 4 | 356 | |||
| HQ830199.1 | - | 4 | 378 | |||

BRSK1
Genomic Map