siRNA Details
| Gene Symbol: JAGN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032492.3 | NP_115881.3 | 2 | 283 | ||
| GenBank mRNA | AF212230.1 | - | 2 | 221 | ||
| AK027508.1 | - | 2 | 229 | |||
| AK074760.1 | - | 2 | 282 | |||
| AK225756.1 | - | 2 | 265 | |||
| BC032101.2 | - | 2 | 263 | 4554225 | ||
| BP262753.1 | - | 2 | 298 | |||

JAGN1
Genomic Map