siRNA Details
| Gene Symbol: ZC3H8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032494.2 | NP_115883.2 | 3 | 358 | ||
| XM_011511998.1 | XP_011510300.1 | 3 | 303 | |||
| XM_017005107.1 | XP_016860596.1 | 3 | 311 | |||
| GenBank mRNA | AF334161.1 | - | 3 | 344 | ||
| AK315329.1 | - | 3 | 312 | |||
| BC032001.1 | - | 3 | 296 | 4749794 | ||
| BG773518.1 | - | 3 | 387 | |||

ZC3H8
Genomic Map