siRNA Details
| Gene Symbol: TMEM185A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282302.1 | NP_001269231.1 | 2 | 501 | ||
| NM_032508.3 | NP_115897.1 | 2 | 501 | |||
| GenBank mRNA | AF530473.1 | - | 3 | 480 | ||
| AK096308.1 | - | 2 | 313 | |||
| BC103820.1 | - | 2 | 205 | |||
| BC103821.1 | - | 2 | 205 | 40000939 | ||
| BC103822.1 | - | 2 | 205 | 40000940 | ||
| BG721461.1 | - | 1 | 100 | |||
| BQ224383.1 | - | 2 | 292 | |||
| DA206736.1 | - | 2 | 393 | |||

TMEM185A
Genomic Map