siRNA Details
| Gene Symbol: FAXC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032511.2 | NP_115900.1 | 3 | 798 | ||
| XM_011536188.2 | XP_011534490.1 | 3 | 315 | |||
| XM_011536189.2 | XP_011534491.1 | 3 | 778 | |||
| GenBank mRNA | AK057793.1 | - | 2 | 216 | ||
| AK096480.1 | - | 3 | 561 | |||
| BC004869.2 | - | 3 | 798 | 3834272 | ||
| BC011583.1 | - | 3 | 798 | 4577456 | ||
| BQ883579.1 | - | 2 | 240 | |||

FAXC
Genomic Map