siRNA Details
| Gene Symbol: SLITRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144003.2 | NP_001137475.1 | 5 | 1235 | ||
| NM_001144004.2 | NP_001137476.1 | 5 | 1223 | |||
| NM_001144005.2 | NP_001137477.1 | 5 | 1245 | |||
| NM_001144006.2 | NP_001137478.1 | 3 | 1455 | |||
| NM_001144008.2 | NP_001137480.1 | 3 | 1467 | |||
| NM_001144009.2 | NP_001137481.1 | 2 | 1308 | |||
| NM_001144010.2 | NP_001137482.1 | 3 | 1445 | |||
| NM_032539.4 | NP_115928.1 | 5 | 1718 | |||
| XM_005262342.3 | XP_005262399.1 | 5 | 2320 | |||
| XM_005262343.3 | XP_005262400.1 | 5 | 2330 | |||
| XM_005262344.3 | XP_005262401.1 | 5 | 2308 | |||
| XM_005262345.3 | XP_005262402.1 | 4 | 2171 | |||
| GenBank mRNA | AB058757.1 | - | 1 | 4983 | ||
| AK091015.1 | - | 1 | 836 | |||
| AK096278.1 | - | 5 | 1714 | |||
| AK289732.1 | - | 5 | 1085 | |||
| BC113011.2 | - | 3 | 1309 | 40028691 | ||
| BC113012.2 | - | 3 | 1311 | 40028693 | ||
| CR627412.1 | - | 2 | 2757 | |||

SLITRK2
Genomic Map