siRNA Details
| Gene Symbol: GPR174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032553.1 | NP_115942.1 | 1 | 771 | ||
| GenBank mRNA | AF345567.1 | - | 1 | 771 | ||
| BC104922.1 | - | 1 | 771 | 8143925 | ||
| BC104924.1 | - | 1 | 771 | 8143927 | ||
More Information
Set Membership: |
|

GPR174
Genomic Map