siRNA Details
| Gene Symbol: SPINK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032566.2 | NP_115955.1 | 3 | 240 | ||
| GenBank mRNA | AF268198.1 | - | 3 | 240 | ||
| AK311570.1 | - | 2 | 941 | |||
| AY359072.1 | - | 3 | 227 | |||
| BC109385.1 | - | 3 | 203 | 40013896 | ||
| BC110067.1 | - | 3 | 203 | 40013897 | ||

SPINK7
Genomic Map