siRNA Details
| Gene Symbol: SLC9A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257291.1 | NP_001244220.1 | 5, 6 | 942 | ||
| NM_032591.2 | NP_115980.1 | 5, 6 | 942 | |||
| XM_005272681.4 | XP_005272738.1 | 5, 6 | 932 | |||
| XM_005272682.4 | XP_005272739.1 | 5, 6 | 932 | |||
| XM_006724564.3 | XP_006724627.1 | 5, 6 | 932 | |||
| XM_011543990.2 | XP_011542292.1 | 5, 6 | 932 | |||
| XM_017029905.1 | XP_016885394.1 | 5, 6 | 932 | |||
| XM_017029906.1 | XP_016885395.1 | 5, 6 | 932 | |||
| GenBank mRNA | AF298591.1 | - | 5, 6 | 807 | ||
| AK056599.1 | - | 3, 4 | 267 | |||

SLC9A7
Genomic Map